missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC56 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | CCDC56 |
---|---|
Concentration | 0.13 mg/mL |
Applications | Immunocytochemistry, Western Blot, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16879964
|
Proteintech
20427-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16869964
|
Proteintech
20427-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
CCDC56 | |
Immunocytochemistry, Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q9D2R6, Q9Y2R0 | |
28958, 498000, 52469 | |
CCDC56 Fusion Protein Ag14260 | |
Primary | |
-20°C | |
COA3 |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
CCDC56, HSPC009 | |
COA3 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CCDC56 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech