missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC56 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20427-1-AP-150UL
This item is not returnable.
View return policy
Specifications
CCDC56 | |
Polyclonal | |
Unconjugated | |
COA3 | |
CCDC56, HSPC009 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
28958, 498000, 52469 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9D2R6, Q9Y2R0 | |
COA3 | |
CCDC56 Fusion Protein Ag14260 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CCDC56 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction