missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCL24/Eotaxin 2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22306-1-AP-150UL
This item is not returnable.
View return policy
Description
Eotaxin 2 belongs to the subfamily of small cytokine CC genes. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cysteines. The cytokine encoded by this gene displays chemotactic activity on resting T lymphocytes, a minimal activity on neutrophils, and is negative on monocytes and activated T lymphocytes. The protein is also a strong suppressor of colony formation by a multipotential hematopoietic progenitor cell line.Specifications
CCL24/Eotaxin 2 | |
Polyclonal | |
Unconjugated | |
CCL24 | |
C C motif chemokine 24, CCL24, CK beta 6, Ckb 6, Eotaxin 2, MPIF 2, MPIF2, SCYA24, Small inducible cytokine A24 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
56221, 6369 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.34 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O00175, Q9JKC0 | |
CCL24 | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CCL24/Eotaxin 2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction