missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCR7 (CD197) Mouse anti-Human , BV605, Clone: 2-L1-A, BD Biosciences™
Mouse Monoclonal Antibody
Brand: BD Biosciences 566755
2625.78 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
CCR7 (CD197) Monoclonal antibody specifically detects CCR7 (CD197) in Human samples. It is validated for Flow Cytometry
Specifications
CCR7 (CD197) | |
Monoclonal | |
Brilliant Violet 605 | |
BLR2; CC chemokine receptor 7; CMKBR7; EBI1; EVI1; Epstein-Barr virus induced gene 1; MIP-3 beta receptor | |
Human CCR7 Transfected Cell Line | |
RUO | |
Human | |
IgG1 Îş |
Flow Cytometry | |
2-L1-A | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Mouse | |
25 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BV605 under optimum conditions, and unconjugated antibody and free BD Horizon™ BV605 were removed. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CCR7 (CD197) Mouse anti-Human , BV605, Clone: 2-L1-A, BD Biosciences™ > 25 Tests; BV605
Spot an opportunity for improvement?Share a Content Correction