Learn More
CD25 (IL-2 Receptor α) Mouse anti-Human, BUV563, Clone: 2A3, BD Horizon™
Mouse Monoclonal Antibody
Brand: BD Biosciences 612919
1446.64 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
CD25 (IL-2 Receptor α) Monoclonal antibody specifically detects CD25 (IL-2 Receptor α) in Human samples. It is validated for Flow Cytometry.
Specifications
CD25 (IL-2 Receptor α) | |
Monoclonal | |
Brilliant Ultraviolet 563 | |
25 Tests | |
Primary | |
Human | |
IgG1 κ |
Flow Cytometry | |
2A3 | |
Mouse | |
RUO | |
CD25 (IL-2 Receptor α) | |
Refrigerate (2 to 8°C) |
Product Content Correction
Product Title
CD25 (IL-2 Receptor α) Mouse anti-Human, BUV563, Clone: 2A3, BD Horizon™ > 25 Tests; BUV563
Spot an opportunity for improvement?Share a Content Correction