missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD279 (PD-1) Mouse anti-Human, Brilliant Ultraviolet 737, Clone: EH12.1 (also known as EH12), BD Horizon™
Mouse Monoclonal Antibody
Brand: BD Biosciences 612792
2326.18 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
CD279 (PD-1) Monoclonal antibody specifically detects CD279 (PD-1) in Human samples. It is validated for Flow cytometry.
Specifications
CD279 (PD-1) | |
Monoclonal | |
Brilliant Ultraviolet 737 | |
Mouse | |
Affinity Purified | |
RUO | |
Human | |
IgG1 Îş |
Flow Cytometry | |
EH12.1 (also known as EH12) | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Human PD-1 Recombinant Protein | |
25 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CD279 (PD-1) Mouse anti-Human, Brilliant Ultraviolet 737, Clone: EH12.1 (also known as EH12), BD Horizon™ > 25 tests; BUV737
Spot an opportunity for improvement?Share a Content Correction