missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Biosciences CD279 (PD-1) Mouse anti-Human , PE-CF594, Clone: MIH4, BD Biosciences™
Mouse Monoclonal Antibody
Brand: BD Biosciences 566846
Additional Details : Weight : 0.00020kg
Description
CD279 (PD-1) Monoclonal antibody specifically detects CD279 (PD-1) in Human samples. It is validated for Flow CytometrySpecifications
CD279 (PD-1) | |
Monoclonal | |
PE-CF594 | |
PD1; hPD-1; hPD-l; PDCD1; PDC1; Programmed cell death 1; SLEB2 | |
Human PD-1 Transfected Cell Line | |
RUO | |
Human | |
Mouse IgG1 κ |
Flow Cytometry | |
MIH4 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Mouse | |
25 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ PE-CF594 under optimum conditions, and unconjugated antibody and free PE-CF594 were removed. |