missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD3 Rat anti-Mouse, APC, Clone: 17A2, BD Pharmingen™
Rat Monoclonal Antibody
Brand: BD Biosciences 565643
This item is not returnable.
View return policy
Description
CD3 Monoclonal antibody specifically detects CD3 in Mouse samples. It is validated for Flow cytometry.
Specifications
CD3 | |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat | |
50 μg | |
Cell Biology | |
Mouse | |
Purified |
Flow Cytometry | |
17A2 | |
APC | |
CD3; CD3 epsilon; Cd3e; CD3ε; T3e | |
γδ TCR-positive T-T hybridoma D1 | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to APC under optimum conditions and unconjugated antibody and free APC were removed. | |
IgG2b κ |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CD3 Rat anti-Mouse, APC, Clone: 17A2, BD Pharmingen™ > 50μg; APC
For Reseach Use Only.
Spot an opportunity for improvement?Share a Content Correction