Learn More
IBA Lifesciences CD4 Fab-Strep Product Code.: 17227600
IBA Lifesciences CD4 Fab-Strep
GREENER_CHOICE
Product Code. 17227600 Shop All IBA Lifesciences Products
Human, 50µg
Click to view available options
Species:
Human
Mouse
Unit Size:
50µg
Product Code. 17227600
Brand: IBA Lifesciences 68002150
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Low affinity Fab fragment fused to a Twin-Strep-tag™
  • CD4 Fab-Strep is a recombinantly expressed low affinity Fab fragment fused to a Twin-Strep-tag™
  • Due to its low affinity for the target, an increase in avidity is required to ensure sufficient binding to the cell surface
  • Combining Fab-Streps with a Strep-Tactin™ backbone leads to the formation of complexes that stably bind to cells
  • Fab-Streps were developed for Traceless Affinity Cell Selection (Fab-TACS™), which is based on our Strep-tag™ technology
Greener Choice Claims
  • Environmental benefits include:
    • Non-Toxic
Specifications
Form Lyophilized
For Use With (Application) Fluorescent Cell Staining & Sorting, Magnetic or Affinity Chromatographic Cell Isolation
Species Human
Sufficient For Human CD4
For Research Use Only.
Product Content Correction
Product Title
IBA Lifesciences CD4 Fab-Strep >