Learn More
CD63 Mouse anti-Human, Unconjugated, Clone: 3D4D1, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67605-1-IG-20UL
 View more versions of this product
Product Code. 16880326
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
The CD63 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CD63. This antibody recognizes human antigen. The CD63 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
CD63
Monoclonal
700 μg/mL
Mouse
Primary
Human
Western Blot, Immunohistochemistry, Immunofluorescence
3D4D1
Unconjugated
20 μL
967
IgG1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CD63 Mouse anti-Human, Unconjugated, Clone: 3D4D1, Proteintech > 20 μL