Learn More
CD63 Mouse anti-Human, Unconjugated, Clone: 3D4D1, Proteintech
Mouse Monoclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | CD63 |
---|---|
Clone | 3D4D1 |
Concentration | 700 ÎĽg/mL |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16880326
|
Proteintech
67605-1-IG-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16870326
|
Proteintech
67605-1-IG-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The CD63 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CD63. This antibody recognizes human antigen. The CD63 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
CD63 | |
700 ÎĽg/mL | |
Monoclonal | |
Mouse | |
967 | |
Primary |
3D4D1 | |
Western Blot, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Human | |
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.