Learn More
Proteintech CD63 Mouse anti-Human, Unconjugated, Clone: 3D4D1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67605-1-IG-20UL
Additional Details : Weight : 0.00010kg
Description
The CD63 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CD63. This antibody recognizes human antigen. The CD63 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
CD63 | |
Monoclonal | |
700 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, Immunohistochemistry, Immunofluorescence | |
3D4D1 | |
Unconjugated | |
20 μL | |
967 | |
IgG1 |