Learn More
CDK1 Mouse anti-Human, Unconjugated, Clone: 2A11E4, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67575-1-IG-20UL
 View more versions of this product
Product Code. 16840286
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
The CDK1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CDK1. This antibody recognizes human, mouse, rat antigen. The CDK1 antibody has been validated for the following applications: FC, WB, ELISA analysis.
Specifications
Specifications
CDK1
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, Flow Cytometry
2A11E4
Unconjugated
20 μL
983
IgG2a
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CDK1 Mouse anti-Human, Unconjugated, Clone: 2A11E4, Proteintech > 20 μL