Learn More
CDK1 Mouse anti-Human, Unconjugated, Clone: 2A11E4, Proteintech
Mouse Monoclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | CDK1 |
---|---|
Clone | 2A11E4 |
Concentration | 1000 ÎĽg/mL |
Applications | Western Blot, Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16840286
|
Proteintech
67575-1-IG-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16830286
|
Proteintech
67575-1-IG-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The CDK1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CDK1. This antibody recognizes human, mouse, rat antigen. The CDK1 antibody has been validated for the following applications: FC, WB, ELISA analysis.
Specifications
CDK1 | |
1000 ÎĽg/mL | |
Monoclonal | |
Mouse | |
983 | |
Primary |
2A11E4 | |
Western Blot, Flow Cytometry | |
Unconjugated | |
Human, Rat, Mouse | |
IgG2a |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.