Learn More
CDK1 Mouse anti-Human, Unconjugated, Clone: 2A11E4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67575-1-IG-150UL
Description
The CDK1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CDK1. This antibody recognizes human, mouse, rat antigen. The CDK1 antibody has been validated for the following applications: FC, WB, ELISA analysis.
Specifications
CDK1 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Rat, Mouse |
Western Blot, Flow Cytometry | |
2A11E4 | |
Unconjugated | |
150 μL | |
983 | |
IgG2a |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.