Learn More
CDK1 Mouse anti-Human, Unconjugated, Clone: 2A11E4, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67575-1-IG-20UL
 View more versions of this product
Product Code. 16840286
  • 1645.54 SEK / 20µL
Description
The CDK1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CDK1. This antibody recognizes human, mouse, rat antigen. The CDK1 antibody has been validated for the following applications: FC, WB, ELISA analysis.
Specifications
Specifications
CDK1
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, Flow Cytometry
2A11E4
Unconjugated
20 μL
983
IgG2a
Certificates
Promotions
Promotions
Product Content Correction
Product Title
CDK1 Mouse anti-Human, Unconjugated, Clone: 2A11E4, Proteintech > 20 μL