Learn More
CDKL5 Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12973-1-AP-150UL
 View more versions of this product
Product Code. 16880954
  • 4652.90 SEK / 150µL
Description
Cyclin dependent kinase like 5 (CDKL5) may regulate ciliogenesis and neuronal morphogenesis. It has been reported that CDKL5 is associated with epileptic seizures, severe intellectual disability, and autistic features. 12973-1-AP antibody detects both two isoforms of CDKL5, separately named as CDKL5a and CDKL5b, around 130-140 and 100-114 kDa in SDS-PAGE.(PMID: 20861382, 28674172, 27315173, 28688852)
Specifications
Specifications
CDKL5
Polyclonal
Unconjugated
150 μL
6792
Liquid
Western Blot, Immunoprecipitation, Immunohistochemistry, ELISA
850 μg/mL
Rabbit
Primary
Human, Mouse
Certificates
Promotions
Promotions
Product Content Correction
Product Title
CDKL5 Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech > 150 μL