Learn More
CDKL5 Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | CDKL5 |
---|---|
Concentration | 850 ÎĽg/mL |
Applications | Western Blot, Immunoprecipitation, Immunohistochemistry, ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16890954
|
Proteintech
12973-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16880954
|
Proteintech
12973-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cyclin dependent kinase like 5 (CDKL5) may regulate ciliogenesis and neuronal morphogenesis. It has been reported that CDKL5 is associated with epileptic seizures, severe intellectual disability, and autistic features. 12973-1-AP antibody detects both two isoforms of CDKL5, separately named as CDKL5a and CDKL5b, around 130-140 and 100-114 kDa in SDS-PAGE.(PMID: 20861382, 28674172, 27315173, 28688852)
Specifications
CDKL5 | |
Western Blot, Immunoprecipitation, Immunohistochemistry, ELISA | |
Unconjugated | |
Rabbit | |
6792 |
850 ÎĽg/mL | |
Polyclonal | |
Liquid | |
Human, Mouse | |
Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.