Learn More
CDKL5 Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12973-1-AP-150UL
 View more versions of this product
Product Code. 16880954
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
Cyclin dependent kinase like 5 (CDKL5) may regulate ciliogenesis and neuronal morphogenesis. It has been reported that CDKL5 is associated with epileptic seizures, severe intellectual disability, and autistic features. 12973-1-AP antibody detects both two isoforms of CDKL5, separately named as CDKL5a and CDKL5b, around 130-140 and 100-114 kDa in SDS-PAGE.(PMID: 20861382, 28674172, 27315173, 28688852)
Specifications
Specifications
CDKL5
Polyclonal
Unconjugated
150 μL
6792
Liquid
Western Blot, Immunoprecipitation, Immunohistochemistry, ELISA
850 μg/mL
Rabbit
Primary
Human, Mouse
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CDKL5 Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech > 150 μL