Learn More
CDKL5 Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12973-1-AP-20UL
Description
Cyclin dependent kinase like 5 (CDKL5) may regulate ciliogenesis and neuronal morphogenesis. It has been reported that CDKL5 is associated with epileptic seizures, severe intellectual disability, and autistic features. 12973-1-AP antibody detects both two isoforms of CDKL5, separately named as CDKL5a and CDKL5b, around 130-140 and 100-114 kDa in SDS-PAGE.(PMID: 20861382, 28674172, 27315173, 28688852)
Specifications
CDKL5 | |
Polyclonal | |
Unconjugated | |
20 μL | |
6792 | |
Liquid |
Western Blot, Immunoprecipitation, Immunohistochemistry, ELISA | |
850 μg/mL | |
Rabbit | |
Primary | |
Human, Mouse |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.