missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEP97 Rabbit anti-Canine, Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22050-1-AP-20UL
This item is not returnable.
View return policy
Description
This is a rabbit polyclonal antibody raised against the C terminus of human CEP97.
CEP97 (centrosomal protein of 97 kDa) has been identified as part of a complex with CP110. Together, CP110 and CEP97 have been shown to play a role in the assembly of cilia. Loss of CEP97 or CEP110 promotes cilia formation while forced expression inhibits ciliogenesis.Specifications
CEP97 | |
Polyclonal | |
Unconjugated | |
CEP97 | |
2810403B08Rik, centrosomal protein 97kDa, Centrosomal protein of 97 kDa, CEP97, LRRIQ2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
608540, 79598 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry, Immunofluorescence | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8IW35 | |
CEP97 | |
CEP97 Fusion Protein Ag16995 | |
20 μL | |
Primary | |
Human, Canine | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CEP97 Rabbit anti-Canine, Human, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction