Learn More
CHOP/GADD153 Mouse anti-Human, Clone: 4F3G1, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66741-1-IG-20UL
 View more versions of this product
Product Code. 16888895
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
GADD153 is a small nuclear protein that is capable of dimerizing with transcription factors C/EBP alpha and beta. Once dimerized, this complex inhibits the normal binding and function of C/EBP to classical binding sites. Inversely, the C/EBP GADD153 dimer gains binding activity to other non classical C/EBP stress related targets. Under normal cellular conditions this protein is not expressed in detectable levels, but is highly unregulated during times of cellular/ER stress. Examples of GADD153 inducing stress include: treatment with tunicamycin, nutrient starvation and reducing agents that interfere with the calcium flux across the ER membrane.
Specifications
Show More
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CHOP/GADD153 Mouse anti-Human, Clone: 4F3G1, Proteintech > 20 μL; Unconjugated