Learn More
Sartorius Stedim Biotech™ Chromatography Papers, Sheets, Grade FN 6
Shop All Sartorius Products
Click to view available options
Unit Size:
Pack of 100
Description
- Highly pure paper grade.
- Ideal for blotting and chromatography.
- Ideal for a wide range of absorption applications like those common in the life sciences and diagnostics.
- Non-sterile, bulk packed.
Specifications
Specifications
Description | 100/Pk, 125 g/m2 |
Dimensions (L x W) | 210 mm × 297 mm |
Film Thickness | 0.22 mm |
Format | Sheet |
Grade | FN 6 |
Material | Cotton Linters (> 98 % alpha-cellulose; < 0.04 % ash) |
Product Content Correction
Product Title
Sartorius Stedim Biotech™ Chromatography Papers, Sheets, Grade FN 6 >
Spot an opportunity for improvement?Share a Content Correction