missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chromium(III) Chloride Hexahydrate, MP Biomedicals
313.04 SEK - 1020.00 SEK
Chemical Identifiers
CAS | 10060-12-5 |
---|---|
Molecular Formula | Cl3CrH12O6 |
Molecular Weight (g/mol) | 266.44 |
MDL Number | MFCD00149660 |
InChI Key | LJAOOBNHPFKCDR-UHFFFAOYSA-K |
Synonym | chromium iii chloride hexahydrate, chromic chloride hexahydrate, hexaaquachromium chloride, chromium trichloride hexahydrate, trichlorochromium hexahydrate, chromium chloride hexahydrate, hexaaquachromium iii chloride, chrom iii-chlorid, chromium 3+ chloride hexahydrate, chlorid chromity hexahydrat czech |
PubChem CID | 104957 |
ChEBI | CHEBI:53442 |
IUPAC Name | chromium(3+) hexahydrate trichloride |
SMILES | O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cr+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11470582
|
MP Biomedicals
0219403380 |
100 g |
313.04 SEK
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11480582
|
MP Biomedicals
0219403390 |
500 g |
1020.00 SEK
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
10060-12-5 | |
266.44 | |
LJAOOBNHPFKCDR-UHFFFAOYSA-K | |
104957 | |
chromium(3+) hexahydrate trichloride |
Cl3CrH12O6 | |
MFCD00149660 | |
chromium iii chloride hexahydrate, chromic chloride hexahydrate, hexaaquachromium chloride, chromium trichloride hexahydrate, trichlorochromium hexahydrate, chromium chloride hexahydrate, hexaaquachromium iii chloride, chrom iii-chlorid, chromium 3+ chloride hexahydrate, chlorid chromity hexahydrat czech | |
CHEBI:53442 | |
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cr+3] |
Specifications
10060-12-5 | |
MFCD00149660 | |
LJAOOBNHPFKCDR-UHFFFAOYSA-K | |
chromium(3+) hexahydrate trichloride | |
104957 | |
266.4 |
Cl3CrH12O6 | |
chromium iii chloride hexahydrate, chromic chloride hexahydrate, hexaaquachromium chloride, chromium trichloride hexahydrate, trichlorochromium hexahydrate, chromium chloride hexahydrate, hexaaquachromium iii chloride, chrom iii-chlorid, chromium 3+ chloride hexahydrate, chlorid chromity hexahydrat czech | |
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cr+3] | |
266.44 | |
CHEBI:53442 | |
Chromium(III) Chloride Hexahydrate |
Safety and Handling
Recommended Storage : Room Temperature
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Chromium(III) Chloride Hexahydrate, MP Biomedicals