Learn More
Thermo Scientific Chemicals Citrate, 0.5M buffer soln., pH 3.0
CAS: 6132-04-3294.10 g/mol
1190.00 SEK - 1695.00 SEK
Chemical Identifiers
CAS 6132-04-3
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15423999
SDS View Documents Promotion Details
Thermo Scientific Chemicals
J61391.AK
250 mL
1190.00 SEK
250mL
15433999
SDS View Documents
Thermo Scientific Chemicals
J61391.AP
500 mL
1695.00 SEK
500mL
Description
Description
Citrate is an intermediate in the TCA cycle and fatty acid synthesis. It is an allosteric modulator of acetyl-CoA carboxylase, the enzyme that regulates the conversion of acetyl-CoA to malonyl-CoA. It has anticoagulant activity and is a calcium chelator. Its function is to break protein cross-links; thus, unmasking antigens and epitopes in formalin-fixed and paraffin embedded tissue sections, resulting in enhancing staining intensity of antibodies.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Citrate is an intermediate in the TCA cycle and fatty acid synthesis. It is an allosteric modulator of acetyl-CoA carboxylase, the enzyme that regulates the conversion of acetyl-CoA to malonyl-CoA. It has anticoagulant activity and is a calcium chelator. Its function is to break protein cross-links; thus, unmasking antigens and epitopes in formalin-fixed and paraffin embedded tissue sections, resulting in enhancing staining intensity of antibodies.
Solubility
Fully miscible in water. Insoluble in ethanol.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
Specifications
Chemical Identifiers
6132-04-3
Specifications
Colorless
294.10
6132-04-3
Keep cold
0.5M buffer soln.
3.0
Odorless
Liquid
Documents
Product Certifications
RUO – Research Use Only
Product Content Correction
Product Title
Thermo Scientific Chemicals Citrate, 0.5M buffer soln., pH 3.0