missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Citrate, 0.5M buffer soln., pH 4.5

CAS: 6132-04-3294.10 g/mol

1090.00 SEK - 1670.00 SEK

Chemical Identifiers

CAS 6132-04-3
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15472869
SDS View Documents Promotion Details
Thermo Scientific Chemicals
J60024.AK
250 mL
1090.00 SEK
250mL
Please to purchase this item. Need a web account? Register with us today!
15482869
SDS View Documents
Thermo Scientific Chemicals
J60024.AP
500 mL
1670.00 SEK
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Citric acid is used as a substrate for citrate lyase, a buffer component, an anticoagulant. For anticoagulation use it is typically used at a concentration of approximately 0.129 M.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Citric acid is used as a substrate for citrate lyase, a buffer component, an anticoagulant. For anticoagulation use it is typically used at a concentration of approximately 0.129 M.

Solubility
Fully miscible with water. Insoluble in ethanol.

Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Chemical Identifiers

6132-04-3

Specifications

Colorless
294.10
6132-04-3
Keep cold
0.5M buffer soln.
4.5
Odorless
Liquid
Videos
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Citrate, 0.5M buffer soln., pH 4.5

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.