Learn More
CLK1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 20439-1-AP-20UL
 View more versions of this product
Product Code. 16879974
  • 1645.54 SEK / 20µL
Description
This gene encodes a member of the CDC2-like (or LAMMER) family of dual specificity protein kinases. In the nucleus, the encoded protein phosphorylates serine/arginine-rich proteins involved in pre-mRNA processing, releasing them into the nucleoplasm. The choice of splice sites during pre-mRNA processing may be regulated by the concentration of transacting factors, including serine/arginine rich proteins. Therefore, the encoded protein may play an indirect role in governing splice site selection.
Specifications
Specifications
CLK1
Polyclonal
Unconjugated
CLK1
CDC like kinase 1, CLK, CLK/STY, CLK1, STY
Rabbit
Antigen Affinity Chromatography
RUO
1195, 12747, 301434
-20°C
Liquid
Immunoprecipitation, Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P22518, P49759
CLK1
CLK1 Fusion Protein Ag14214
20 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Certificates
Promotions
Promotions
Product Content Correction
Product Title
CLK1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated