Learn More
CLTCL1 Mouse anti-Human, Clone: 1B5A10, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66274-1-IG-20UL
 View more versions of this product
Product Code. 16887995
  • 1645.54 SEK / 20µL
Description
The antibody is specific to CLTCL1. It has no cross reaction to CLTC.
Clathrin is the major protein of the polyhedral coat of coated pits and vesicles. Two different adapter protein complexes link the clathrin lattice either to the plasma membrane or to the trans-Golgi network.
Specifications
Show More
Certificates
Promotions
Promotions
Product Content Correction
Product Title
CLTCL1 Mouse anti-Human, Clone: 1B5A10, Proteintech > 20 μL; Unconjugated