missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLTCL1 Mouse anti-Human, Clone: 1B5A10, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66274-1-IG-150UL
This item is not returnable.
View return policy
Description
The antibody is specific to CLTCL1. It has no cross reaction to CLTC.
Clathrin is the major protein of the polyhedral coat of coated pits and vesicles. Two different adapter protein complexes link the clathrin lattice either to the plasma membrane or to the trans-Golgi network.Specifications
CLTCL1 | |
Monoclonal | |
1.2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P53675 | |
CLTCL1 | |
Peptide | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1B5A10 | |
Unconjugated | |
CLTCL1 | |
CHC22, Clathrin heavy chain 2, clathrin, heavy chain like 1, CLH 22, CLH22, CLTCL, CLTCL1, CLTD | |
Mouse | |
Protein A | |
RUO | |
8218 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CLTCL1 Mouse anti-Human, Clone: 1B5A10, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction