Learn More
Eppendorf™ Conical Bottom 384 Well Twin Tec PCR Plate
2745.00 SEK - 30421.20 SEK
Specifications
Format | Standard |
---|---|
Array | 16 x 24 |
No. of Wells | 384 |
Well Shape | Conical Bottom (V-well) |
Product Code | Brand | Color | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | ||||||
10529113
![]() |
Eppendorf™
0030128893 |
Black, Clear Bottom |
30421.20 SEK
Pack of 300 |
Estimated Shipment: 13-08-2025
Log in to see stock. |
||||||
10724977
![]() |
Eppendorf™
0030128818 |
Black, Yellow | N/A | |||||||
10218354
![]() |
Eppendorf™
0030128966 |
Blue |
28112.00 SEK
Pack of 300 |
In Stock
Log in to see stock. |
||||||
10764767
![]() |
Eppendorf™
0030128958 |
Green |
28112.00 SEK
Pack of 300 |
In Stock
Log in to see stock. |
||||||
10052043
![]() |
Eppendorf™
0030128885 |
Orange, Clear Bottom |
29015.60 SEK
Pack of 300 |
In Stock
Log in to see stock. |
||||||
10238744
![]() |
Eppendorf™
0030128974 |
Red |
28112.00 SEK
Pack of 300 |
In Stock
Log in to see stock. |
||||||
10075384
![]() |
Eppendorf™
0030128940 |
Yellow |
28112.00 SEK
Pack of 300 |
In Stock
Log in to see stock. |
||||||
Description
Specifications
Standard | |
16 x 24 | |
384 | |
Conical Bottom (V-well) |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Eppendorf™ Conical Bottom 384 Well Twin Tec PCR Plate