missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COSMC Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19254-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a type II transmembrane protein that is similar to the core 1 beta1,3-galactosyltransferase 1, which catalyzes the synthesis of the core-1 structure, also known as Thomsen-Friedenreich antigen, on O-linked glycans. This gene product lacks the galactosyltransferase activity itself, but instead acts as a molecular chaperone required for the folding, stability and full activity of the core 1 beta1,3-galactosyltransferase 1. Mutations in this gene have been associated with Tn syndrome. Alternatively spliced transcript variants encoding the same protein have been identified.Specifications
COSMC | |
Polyclonal | |
Unconjugated | |
C1GALT1C1 | |
C1Gal T2, C1GALT1 chaperone 1, C1GALT1 specific chaperone 1, C1GALT1C1, C1GALT2, c38h2 l1, C38H2 like protein 1, Core 1 beta3 Gal T2, COSMC, HSPC067, MSTP143, UNQ273/PRO310 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
29071, 302499, 59048 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q499P3, Q96EU7, Q9JMG2 | |
C1galt1c1 | |
COSMC Fusion Protein Ag6899 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
COSMC Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction