Learn More
Hirschmann™ Counting Chambers Double Net Ruling
Facilitates quick and easy determination of cell concentration in liquid samples. Hirschmann™ Counting Chambers are available in a variety of depths to fit a range of systems.
Brand: Hirschmann™ 8100101
714.96 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Double net ruling
- Including 2 cover glasses for hemacytometer
- CE acc. IVD 98/79 EC
Certifications
CE acc. IVD 98/79 EC
Specifications
Counting chambers, double net ruling | |
1 Each | |
2 cover glasses for haemacytometer |
Counting chamber acc. to Bürker | |
0.1 mm |
Product Content Correction
Product Title
Hirschmann™ Counting Chambers Double Net Ruling > Description: Counting chamber acc. to Bürker
Spot an opportunity for improvement?Share a Content Correction