Learn More
Hirschmann™ Counting Chambers Double Net Ruling
Facilitates quick and easy determination of cell concentration in liquid samples. Hirschmann™ Counting Chambers are available in a variety of depths to fit a range of systems.
724.00 SEK - 2445.00 SEK
Specifications
Product Type | Counting chambers, double net ruling |
---|---|
Quantity | 1 Each |
Includes | 2 cover glasses for haemacytometer |
Product Code | Brand | Description | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | ||||||
10638902
|
Hirschmann™
8100105 |
Counting chamber acc. to Thoma |
733.00 SEK
Each |
In Stock
Log in to see stock. |
||||||
10448851
|
Hirschmann™
8100106 |
Counting chamber acc. to Thoma - new |
733.00 SEK
Each |
In Stock
Log in to see stock. |
||||||
10521412
|
Hirschmann™
8100111 |
Counting chamber acc. to Nageotte |
2445.00 SEK
Each |
In Stock
Log in to see stock. |
||||||
10438701
|
Hirschmann™
8100101 |
Counting chamber acc. to BĂĽrker |
724.00 SEK
Each |
In Stock
Log in to see stock. |
||||||
Description
- Double net ruling
- Including 2 cover glasses for hemacytometer
- CE acc. IVD 98/79 EC
Certifications
CE acc. IVD 98/79 EC
Specifications
Counting chambers, double net ruling | |
1 Each | |
2 cover glasses for haemacytometer |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Hirschmann™ Counting Chambers Double Net Ruling