Learn More
Hirschmann™ Counting Chambers Double Net Ruling
Facilitates quick and easy determination of cell concentration in liquid samples. Hirschmann™ Counting Chambers are available in a variety of depths to fit a range of systems.
Brand:  Hirschmann™ 8100106
714.96 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10448851
  • 733.00 SEK / Each
Only null left
In Stock
Description
  • Double net ruling
  • Including 2 cover glasses for hemacytometer
  • CE acc. IVD 98/79 EC
Certifications
CE acc. IVD 98/79 EC
Specifications
Specifications
Counting chambers, double net ruling
1 Each
2 cover glasses for haemacytometer
Counting chamber acc. to Thoma - new
0.1 mm
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Hirschmann™ Counting Chambers Double Net Ruling > Description: Counting chamber acc. to Thoma - new