Learn More
CPS1 Mouse anti-Human, Unconjugated, Clone: 2A12A4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67630-1-IG-20UL
Description
The CPS1 antibody from Proteintech is a mouse monoclonal antibody generated with peptide of human CPS1. This antibody recognizes human, mouse, rat antigen. The CPS1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
CPS1 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Rat, Mouse |
Western Blot, ELISA | |
2A12A4 | |
Unconjugated | |
20 μL | |
1373 | |
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.