missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CPT1B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22170-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene, a member of the carnitine/choline acetyltransferase family, is the rate-controlling enzyme of the long-chain fatty acid beta-oxidation pathway in muscle mitochondria. This enzyme is required for the net transport of long-chain fatty acyl-CoAs from the cytoplasm into the mitochondria. Multiple transcript variants encoding different isoforms have been found for this gene, and one of the transcripts is bicistronic.Specifications
CPT1B | |
Polyclonal | |
Unconjugated | |
CPT1B | |
CPT I, CPT1 M, CPT1B, CPTI M, KIAA1670, M CPT1, MCCPT1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
12895, 1375, 25756 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q63704, Q924X2, Q92523 | |
CPT1B | |
CPT1B-specific Fusion Protein Ag17840 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CPT1B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction