Learn More
CREB3L1 OASIS Mouse anti-Human, Unconjugated, Clone: 2B5B4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67617-1-IG-20UL
Description
The CREB3L1, OASIS antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CREB3L1, OASIS. This antibody recognizes human, mouse, rat antigen. The CREB3L1, OASIS antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
CREB3L1 OASIS | |
Monoclonal | |
1000 ÎĽg/mL | |
Mouse | |
Primary | |
Human, Rat, Mouse |
Western Blot, ELISA | |
2B5B4 | |
Unconjugated | |
20 ÎĽL | |
90993 | |
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.