missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CREST Mouse anti-Human, Mouse, Rat, Clone: 3D5D10, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 60314-1-IG-20UL
This item is not returnable.
View return policy
Description
Synovial sarcomas occur most frequently in the extremities around large joints. More than 90% of cases have a recurrent and specific chromosomal translocation, t(X;18)(p11.2;q11.2), in which the 5-prime end of the SS18 gene (MIM 600192) is fused in-frame to the 3-prime end of the SSX1 (MIM 312820), SSX2 (MIM 300192), or SSX4 (MIM 300326) gene. The SS18L1 gene is homologous to SS18.Specifications
CREST | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O75177, Q8BW22, Q91XJ0 | |
Ss18l1 | |
CREST Fusion Protein Ag3119 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG1 |
Western Blot | |
3D5D10 | |
Unconjugated | |
SS18L1 | |
CREST, KIAA0693, LP2261, SS18 like protein 1, SS18L1, SYT homolog 1 | |
Mouse | |
Protein G | |
RUO | |
192352, 26039, 269397 | |
-20°C | |
Liquid |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CREST Mouse anti-Human, Mouse, Rat, Clone: 3D5D10, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction