Learn More
CRH/CRF Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10944-1-AP-150UL
 View more versions of this product
Product Code. 16877663
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
This item is not returnable. View return policy
Description
Description
The CRH/CRF antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CRH/CRF. This antibody recognizes human, mouse, rat antigen. The CRH/CRF antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
CRH/CRF
Polyclonal
Unconjugated
150 μL
1392
IgG
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
400 μg/mL
Rabbit
Primary
Human, Rat, Mouse
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CRH/CRF Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech > 150 μL