Learn More
CRH/CRF Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | CRH/CRF |
---|---|
Concentration | 400 ÎĽg/mL |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence, ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
The CRH/CRF antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CRH/CRF. This antibody recognizes human, mouse, rat antigen. The CRH/CRF antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
CRH/CRF | |
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
Unconjugated | |
Human, Rat, Mouse | |
IgG |
400 ÎĽg/mL | |
Polyclonal | |
Rabbit | |
1392 | |
Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.