missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRLS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14845-1-AP-20UL
This item is not returnable.
View return policy
Description
The function of this protein is still unknown. The gene of human hypothetical protein (LOC54675) is up-regulated in colon cancer metastasis.Specifications
CRLS1 | |
Polyclonal | |
Unconjugated | |
CRLS1 | |
C20orf155, Cardiolipin synthase, cardiolipin synthase 1, CLS, CLS1, CRLS1, GCD10, Protein GCD10 homolog | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
366196, 54675, 66586 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q5U2V5, Q80ZM8, Q9UJA2 | |
Crls1 | |
Peptide | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CRLS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction