missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRYBB3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21009-1-AP-150UL
This item is not returnable.
View return policy
Description
Crystallins are the domit structural components of the vertebrate eye lens.Specifications
CRYBB3 | |
Polyclonal | |
Unconjugated | |
CRYBB3 | |
Beta B3 crystallin, Beta crystallin B3, CATCN2, CRYB3, CRYBB3, crystallin, beta B3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
12962, 1417, 64349 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.49 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P02524, P26998, Q9JJU9 | |
Crybb3 | |
CRYBB3 Fusion Protein Ag15212 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CRYBB3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction