missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Biosciences CX3CR1 Rat anti-Human , PE-CF594, Clone: 2A9-1, BD Biosciences™
Rat Monoclonal Antibody
Brand: BD Biosciences 565897
Additional Details : Weight : 0.00060kg
Description
CX3CR1 Monoclonal antibody specifically detects CX3CR1 in Human samples. It is validated for Flow CytometrySpecifications
CX3CR1 | |
Monoclonal | |
PE-CF594 | |
CCRL1; CMKBRL1; CMKDR1; GPR13; GPRV28; V28; Fractalkine Receptor | |
Human CX3CR1 Recombinant Protein | |
RUO | |
Human | |
Rat IgG2b κ |
Flow Cytometry | |
2A9-1 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Rat | |
100 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ PE-CF594 under optimum conditions, and unconjugated antibody and free PE-CF594 were removed. |