Learn More
CXCL16 Mouse anti-Human, Mouse, Clone: 2H7B3, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 60123-1-IG-20UL
 View more versions of this product
Product Code. 16887085
  • 1662.46 SEK / 20µL
Description
CXCL16 acts as a scavenger receptor on macrophages, which specifically binds to OxLDL (oxidized low density lipoprotein), suggesting that it may be involved in pathophysiology such as atherogenesis. Induces a strong chemotactic response. Induces calcium mobilization. Binds to CXCR6/Bonzo.
Specifications
Show More
Certificates
Promotions
Promotions
Product Content Correction
Product Title
CXCL16 Mouse anti-Human, Mouse, Clone: 2H7B3, Proteintech > 20 μL; Unconjugated