missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCL16 Mouse anti-Human, Mouse, Clone: 2H7B3, Proteintech
Mouse Monoclonal Antibody
1662.46 SEK - 4700.76 SEK
Specifications
Antigen | CXCL16 |
---|---|
Clone | 2H7B3 |
Concentration | 1.33 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16887085
|
Proteintech
60123-1-IG-20UL |
20 ÎĽL |
1662.46 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16877085
|
Proteintech
60123-1-IG-150UL |
150 ÎĽL |
4700.76 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CXCL16 acts as a scavenger receptor on macrophages, which specifically binds to OxLDL (oxidized low density lipoprotein), suggesting that it may be involved in pathophysiology such as atherogenesis. Induces a strong chemotactic response. Induces calcium mobilization. Binds to CXCR6/Bonzo.Specifications
CXCL16 | |
1.33 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
C X C motif chemokine 16, CXCL16, CXCLG16, SCYB16, Small inducible cytokine B16, SR PSOX, SRPSOX, Transmembrane chemokine CXCL16 | |
CXCL16 | |
IgG2a | |
Caprylic Acid Precipitation | |
Antibody |
2H7B3 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse | |
Q8BSU2, Q9H2A7 | |
58191, 66102 | |
CXCL16 Fusion Protein Ag4883 | |
Primary | |
-20°C | |
CXCL16 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CXCL16 Mouse anti-Human, Mouse, Clone: 2H7B3, Proteintech