Learn More
CYP1B1 Mouse anti-Human, Unconjugated, Clone: 1G8D4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67033-1-IG-150UL
Description
The CYP1B1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CYP1B1. This antibody recognizes human, mouse, rat, pig antigen. The CYP1B1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
CYP1B1 | |
Monoclonal | |
500 μg/mL | |
Mouse | |
Primary | |
Human, Mouse, Rat, Pig |
Western Blot, ELISA | |
1G8D4 | |
Unconjugated | |
150 μL | |
1545 | |
IgG1 |