Learn More
DAAM1 Mouse anti-Human, Unconjugated, Clone: 3F11F11, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67287-1-IG-150UL
 View more versions of this product
Product Code. 16879825
  • 4652.90 SEK / 150µL
Explore more special offers
Description
Description
The DAAM1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human DAAM1. This antibody recognizes human, mouse, rat antigen. The DAAM1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
DAAM1
Monoclonal
920 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, ELISA
3F11F11
Unconjugated
150 μL
23002
IgG1
Product Title
DAAM1 Mouse anti-Human, Unconjugated, Clone: 3F11F11, Proteintech > 150 μL