Learn More
DAAM1 Mouse anti-Human, Unconjugated, Clone: 3F11F11, Proteintech
Mouse Monoclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | DAAM1 |
---|---|
Clone | 3F11F11 |
Concentration | 920 ÎĽg/mL |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16889825
|
Proteintech
67287-1-IG-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16879825
|
Proteintech
67287-1-IG-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The DAAM1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human DAAM1. This antibody recognizes human, mouse, rat antigen. The DAAM1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
DAAM1 | |
920 ÎĽg/mL | |
Monoclonal | |
Mouse | |
23002 | |
Primary |
3F11F11 | |
Western Blot, ELISA | |
Unconjugated | |
Human, Rat, Mouse | |
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.