Learn More
DAAM1 Mouse anti-Human, Unconjugated, Clone: 3F11F11, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67287-1-IG-20UL
 View more versions of this product
Product Code. 16889825
  • 1645.54 SEK / 20µL
Description
The DAAM1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human DAAM1. This antibody recognizes human, mouse, rat antigen. The DAAM1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
DAAM1
Monoclonal
920 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, ELISA
3F11F11
Unconjugated
20 μL
23002
IgG1
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
DAAM1 Mouse anti-Human, Unconjugated, Clone: 3F11F11, Proteintech > 20 μL