Learn More
DCDC2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 26978-1-AP-20UL
 View more versions of this product
Product Code. 16885295
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
This item is not returnable. View return policy
Description
Description
This gene encodes a member of the doublecortin family. The protein encoded by this gene contains two doublecortin domains. The doublecortin domain has been demonstrated to bind tubulin and enhance microtubule polymerization. Mutations in this gene have been associated with Reading Disability, also referred to as developmental dyslexia. Alternatively spliced transcript variants encoding the same protein have been found for this gene.
Specifications
Specifications
DCDC2
Polyclonal
Unconjugated
DCDC2
DCDC2, DCDC2A, KIAA1154, Protein RU2S, RU2, RU2S
Rabbit
Antigen Affinity Chromatography
RUO
195208, 51473
-20°C
Liquid
Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry
0.33 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q5DU00, Q9UHG0
DCDC2
DCDC2 Fusion Protein Ag25577
20 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DCDC2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated